siRNA Details
| Gene Symbol: Zbtb9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213564.3 | NP_998729.1 | 2 | 1757 | ||
| XM_006256121.2 | XP_006256183.1 | 2 | 1745 | |||
| XM_006256122.2 | XP_006256184.1 | 2 | 1690 | |||
| GenBank mRNA | BC100649.1 | - | 1705 | 7932768 | ||

Zbtb9
Genomic Map