siRNA Details
| Gene Symbol: Mrfap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009264.1 | NP_001009264.1 | 2 | 695 | ||
| GenBank mRNA | BC088308.1 | - | 2 | 695 | 7324285 | |
| FQ228750.1 | - | 2 | 715 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrfap1| Gene Symbol: Mrfap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009264.1 | NP_001009264.1 | 2 | 695 | ||
| GenBank mRNA | BC088308.1 | - | 2 | 695 | 7324285 | |
| FQ228750.1 | - | 2 | 715 | |||