siRNA Details
| Gene Symbol: Nubp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009619.1 | NP_001009619.1 | 11 | 1169 | ||
| GenBank mRNA | BC088221.1 | - | 11 | 1169 | 7321552 | |
| FQ214074.1 | - | 10 | 1113 | |||
| FQ218444.1 | - | 11 | 1182 | |||
| FQ222868.1 | - | 11 | 1186 | |||
| FQ229330.1 | - | 11 | 1185 | |||

Nubp1
Genomic Map