siRNA Details
| Gene Symbol: Tmem204 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009620.1 | NP_001009620.1 | 3 | 1466 | ||
| GenBank mRNA | BC087601.1 | - | 3 | 1466 | 7314703 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem204| Gene Symbol: Tmem204 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009620.1 | NP_001009620.1 | 3 | 1466 | ||
| GenBank mRNA | BC087601.1 | - | 3 | 1466 | 7314703 | |