siRNA Details
| Gene Symbol: Fam175a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009633.1 | NP_001009633.1 | 5 | 466 | ||
| GenBank mRNA | BC088408.1 | - | 5 | 466 | 7131771 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam175a| Gene Symbol: Fam175a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009633.1 | NP_001009633.1 | 5 | 466 | ||
| GenBank mRNA | BC088408.1 | - | 5 | 466 | 7131771 | |