siRNA Details
| Gene Symbol: Spryd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009635.1 | NP_001009635.1 | 5 | 596 | ||
| GenBank mRNA | BC088471.1 | - | 5 | 596 | 7128422 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spryd7| Gene Symbol: Spryd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009635.1 | NP_001009635.1 | 5 | 596 | ||
| GenBank mRNA | BC088471.1 | - | 5 | 596 | 7128422 | |