siRNA Details
| Gene Symbol: VPS13C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001018088.2 | NP_001018098.1 | 74 | 10132 | ||
| NM_017684.4 | NP_060154.3 | 72 | 10003 | |||
| NM_018080.3 | NP_060550.2 | 72 | 10003 | |||
| NM_020821.2 | NP_065872.1 | 74 | 10132 | |||
| GenBank mRNA | AJ608770.1 | - | 72 | 9986 | ||
| AJ608771.1 | - | 74 | 10115 | |||
| AJ626860.1 | - | 72 | 9986 | |||
| AJ626861.1 | - | 74 | 10115 | |||
| AK000143.1 | - | 2 | 131 | |||
| AK123851.1 | - | 7 | 690 | |||
| AK128650.1 | - | 28 | 4546 | |||
| AK304408.1 | - | 9 | 1037 | |||
| AL833377.1 | - | 8 | 733 | |||
| BC069387.1 | - | 2 | 131 | |||
| BX648766.1 | - | 10 | 1084 | |||
| CR936716.1 | - | 23 | 3614 | |||

VPS13C
Genomic Map