siRNA Details
| Gene Symbol: Zgpat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009656.1 | NP_001009656.1 | 2 | 775 | ||
| XM_006235749.1 | XP_006235811.1 | 3 | 771 | |||
| XM_006235750.1 | XP_006235812.1 | 3 | 736 | |||
| XM_006235751.2 | XP_006235813.1 | 3 | 757 | |||
| XM_006235752.2 | XP_006235814.1 | 2 | 671 | |||
| GenBank mRNA | BC087720.1 | - | 2 | 775 | 7318724 | |

Zgpat
Genomic Map