siRNA Details
| Gene Symbol: Thnsl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009658.1 | NP_001009658.1 | 4 | 567 | ||
| XM_006236625.2 | XP_006236687.1 | 697 | ||||
| XM_006236626.2 | XP_006236688.1 | 697 | ||||
| XM_006236627.2 | XP_006236689.1 | 503 | ||||
| GenBank mRNA | BC088454.1 | - | 4 | 567 | 7127050 | |

Thnsl2
Genomic Map