siRNA Details
| Gene Symbol: Thnsl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009658.1 | NP_001009658.1 | 7, 8 | 1130 | ||
| XM_006236625.2 | XP_006236687.1 | 1260 | ||||
| XM_006236626.2 | XP_006236688.1 | 1260 | ||||
| XM_006236627.2 | XP_006236689.1 | 1066 | ||||
| GenBank mRNA | BC088454.1 | - | 7, 8 | 1130 | 7127050 | |

Thnsl2
Genomic Map