siRNA Details
| Gene Symbol: Ccdc174 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009659.2 | NP_001009659.2 | 4, 5 | 375 | ||
| XM_006236907.2 | XP_006236969.1 | 4, 5 | 412 | |||
| GenBank mRNA | BC087050.1 | - | 4, 5 | 375 | 7114831 | |

Ccdc174
Genomic Map