siRNA Details
| Gene Symbol: Uba5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009669.1 | NP_001009669.1 | 8 | 859 | ||
| GenBank mRNA | BC088757.1 | - | 8 | 859 | 7309521 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Uba5| Gene Symbol: Uba5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009669.1 | NP_001009669.1 | 8 | 859 | ||
| GenBank mRNA | BC088757.1 | - | 8 | 859 | 7309521 | |