siRNA Details
| Gene Symbol: Coq10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009671.1 | NP_001009671.1 | 5 | 866 | ||
| XM_008767080.1 | XP_008765302.1 | 5 | 1167 | |||
| GenBank mRNA | BC088273.1 | - | 5 | 866 | 7380093 | |
| FQ219530.1 | - | 4 | 725 | |||

Coq10b
Genomic Map