siRNA Details
Gene Symbol: BIVM-ERCC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204425.1 | NP_001191354.1 | 1 | 221 | ||
GenBank mRNA | AB209312.1 | - | 1 | 13 | ||
LM994822.1 | - | 1 | 99 |
Gene Symbol: BIVM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017693.3 | NP_060163.2 | 3 | 778 | ||
GenBank mRNA | AF411385.1 | - | 3 | 778 | ||
AK290658.1 | - | 3 | 647 | |||
AK304391.1 | - | 2 | 532 | |||
BC075084.2 | - | 1 | 126 | 30915598 | ||
BC075085.2 | - | 1 | 126 | 30915386 |
