siRNA Details
| Gene Symbol: Tceal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009675.1 | NP_001009675.1 | 2 | 441 | ||
| XM_006257294.1 | XP_006257356.1 | 2 | 506 | |||
| XM_008773413.1 | XP_008771635.1 | 2 | 594 | |||
| XM_008773414.1 | XP_008771636.1 | 2 | 455 | |||
| GenBank mRNA | BC087575.1 | - | 2 | 441 | 7302183 | |
| FQ214955.1 | - | 2 | 374 | |||

Tceal1
Genomic Map