siRNA Details
| Gene Symbol: Ppp1r7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009825.1 | NP_001009825.1 | 10 | 2279 | ||
| GenBank mRNA | BC088868.1 | - | 10 | 2279 | 7321115 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppp1r7| Gene Symbol: Ppp1r7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009825.1 | NP_001009825.1 | 10 | 2279 | ||
| GenBank mRNA | BC088868.1 | - | 10 | 2279 | 7321115 | |