siRNA Details
| Gene Symbol: FAM102B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010883.2 | NP_001010883.2 | 7, 8 | 740 | ||
| GenBank mRNA | BC127898.1 | - | 7, 8 | 740 | 40132918 | |
| BC140863.1 | - | 7, 8 | 596 | 9021731 | ||
| BC140865.1 | - | 7, 8 | 596 | 9021733 | ||
| BC150530.1 | - | 7, 8 | 740 | 40132919 | ||
| CR749397.1 | - | 4, 5 | 311 | |||
| CR936815.1 | - | 1 | 3724 | |||

FAM102B
Genomic Map