siRNA Details
| Gene Symbol: Tmem120a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010945.1 | NP_001010945.1 | 3 | 622 | ||
| GenBank mRNA | BC089063.1 | - | 3 | 622 | 7310299 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem120a| Gene Symbol: Tmem120a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010945.1 | NP_001010945.1 | 3 | 622 | ||
| GenBank mRNA | BC089063.1 | - | 3 | 622 | 7310299 | |