siRNA Details
| Gene Symbol: Tmem126a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011557.1 | NP_001011557.1 | 3, 4 | 364 | ||
| XM_006229689.2 | XP_006229751.1 | 3, 4 | 529 | |||
| GenBank mRNA | BC089104.1 | - | 3, 4 | 364 | 7307750 | |
| FQ210414.1 | - | 3, 4 | 385 | |||
| FQ220582.1 | - | 3, 4 | 523 | |||
| FQ231598.1 | - | 3, 4 | 414 | |||

Tmem126a
Genomic Map