siRNA Details
| Gene Symbol: Tmem126a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011557.1 | NP_001011557.1 | 5 | 1470 | ||
| GenBank mRNA | BC089104.1 | - | 5 | 1470 | 7307750 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem126a| Gene Symbol: Tmem126a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011557.1 | NP_001011557.1 | 5 | 1470 | ||
| GenBank mRNA | BC089104.1 | - | 5 | 1470 | 7307750 | |