siRNA Details
| Gene Symbol: Trpm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011559.1 | NP_001011559.1 | 27 | 3885 | ||
| XM_006256248.2 | XP_006256310.1 | 28 | 4075 | |||
| XM_006256249.2 | XP_006256311.1 | 28 | 4072 | |||
| XM_008772811.1 | XP_008771033.1 | 28 | 4123 | |||
| GenBank mRNA | AB193179.1 | - | 27 | 3913 | ||
| AY749166.1 | - | 27 | 3885 | |||

Trpm2
Genomic Map