siRNA Details
| Gene Symbol: Olfr685 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011857.1 | NP_001011857.1 | 1 | 432 | ||
| GenBank mRNA | BC147196.1 | - | 1 | 521 | 9056252 | |
| BC147197.1 | - | 1 | 521 | 9056258 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr685| Gene Symbol: Olfr685 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011857.1 | NP_001011857.1 | 1 | 432 | ||
| GenBank mRNA | BC147196.1 | - | 1 | 521 | 9056252 | |
| BC147197.1 | - | 1 | 521 | 9056258 | ||