siRNA Details
| Gene Symbol: Serpinf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011892.1 | NP_001011892.1 | 10 | 2150 | ||
| XM_006246675.1 | XP_006246737.1 | 10 | 2295 | |||
| GenBank mRNA | BC079362.1 | - | 10 | 2150 | 7109243 | |
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_008878, AA269351, BC026756, AK090208, AK077563, and others.

Serpinf2
Genomic Map