siRNA Details
| Gene Symbol: Sept4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011893.1 | NP_001011893.1 | 6, 7 | 718 | ||
| XM_008767966.1 | XP_008766188.1 | 10, 11 | 1898 | |||
| GenBank mRNA | AY208293.1 | - | 6, 7 | 718 | ||
| AY208294.1 | - | 6, 7 | 718 | |||
| BC088334.1 | - | 5, 6 | 566 | |||
| BC126069.1 | - | 6, 7 | 619 | 7441949 | ||

Sept4
Genomic Map