siRNA Details
| Gene Symbol: Ndufs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011907.1 | NP_001011907.1 | 5 | 549 | ||
| XM_003751277.2 | XP_003751325.1 | 5 | 562 | |||
| GenBank mRNA | BC082067.1 | - | 5 | 549 | 7132777 | |
| FQ215922.1 | - | 5 | 553 | |||
| Gene Symbol: LOC103690046 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769779.1 | XP_008768001.1 | 2 | 180 | ||

Ndufs2
Genomic Map