siRNA Details
| Gene Symbol: ASAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143778.1 | NP_001137250.1 | 1, 2 | 258 | ||
| NM_017707.3 | NP_060177.2 | 1, 2 | 258 | |||
| XM_011541755.1 | XP_011540057.1 | 1, 2 | 192 | |||
| XM_017001685.1 | XP_016857174.1 | 1, 2 | 192 | |||
| XM_017001686.1 | XP_016857175.1 | 1, 2 | 192 | |||
| GenBank mRNA | AB056749.1 | - | 1, 2 | 179 | ||
| AK092291.1 | - | 1, 2 | 180 | |||
| AK295100.1 | - | 1, 2 | 180 | |||
| BC023519.2 | - | 2, 3 | 246 | |||
| BC060786.1 | - | 1, 2 | 206 | 5298355 | ||

ASAP3
Genomic Map