siRNA Details
| Gene Symbol: FAM83E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017708.3 | NP_060178.2 | 3 | 810 | ||
| XM_005259019.1 | XP_005259076.1 | 4 | 2134 | |||
| XM_011527049.2 | XP_011525351.1 | 4 | 2119 | |||
| GenBank mRNA | AK000207.1 | - | 3 | 936 | ||
| BC111970.1 | - | 3 | 810 | 8327438 | ||
| BC111972.1 | - | 3 | 810 | 8327440 | ||

FAM83E
Genomic Map