siRNA Details
| Gene Symbol: Lyar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011911.1 | NP_001011911.1 | 6 | 647 | ||
| XM_006251087.1 | XP_006251149.1 | 6 | 653 | |||
| GenBank mRNA | BC079008.1 | - | 6 | 647 | 7113133 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lyar| Gene Symbol: Lyar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011911.1 | NP_001011911.1 | 6 | 647 | ||
| XM_006251087.1 | XP_006251149.1 | 6 | 653 | |||
| GenBank mRNA | BC079008.1 | - | 6 | 647 | 7113133 | |