siRNA Details
| Gene Symbol: Polm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011912.1 | NP_001011912.1 | 11 | 2404 | ||
| XM_006251211.2 | XP_006251273.1 | 11 | 2618 | |||
| XM_006251212.2 | XP_006251274.1 | 11 | 2606 | |||
| XM_006251213.2 | XP_006251275.1 | 11 | 2784 | |||
| XM_006251214.2 | XP_006251276.1 | 11 | 2026 | |||
| GenBank mRNA | BC081868.1 | - | 11 | 2404 | 7096894 | |

Polm
Genomic Map