siRNA Details
| Gene Symbol: Dr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011914.1 | NP_001011914.1 | 3 | 1022 | ||
| GenBank mRNA | BC083822.1 | - | 3 | 1022 | 7111461 | |
| Gene Symbol: LOC100910207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751317.3 | XP_003751365.1 | 1111 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_026106, AK075713, AK090113, AK144749, AK146573, and others.

Dr1
Genomic Map