siRNA Details
| Gene Symbol: Fermt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011915.1 | NP_001011915.1 | 6 | 935 | ||
| XM_006251755.2 | XP_006251817.1 | 7 | 993 | |||
| XM_006251756.2 | XP_006251818.1 | 7 | 947 | |||
| XM_006251757.2 | XP_006251819.1 | 7 | 993 | |||
| XM_006251758.2 | XP_006251820.1 | 6 | 960 | |||
| XM_006251759.2 | XP_006251821.1 | 7 | 993 | |||
| XM_006251760.2 | XP_006251822.1 | 6 | 960 | |||
| XM_006251761.2 | XP_006251823.1 | 6 | 957 | |||
| GenBank mRNA | BC083876.1 | - | 6 | 935 | 7129898 | |

Fermt2
Genomic Map