siRNA Details
| Gene Symbol: Sugp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011920.1 | NP_001011920.1 | 12, 13 | 1797 | ||
| GenBank mRNA | BC079341.1 | - | 12, 13 | 1797 | 7106652 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sugp1| Gene Symbol: Sugp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011920.1 | NP_001011920.1 | 12, 13 | 1797 | ||
| GenBank mRNA | BC079341.1 | - | 12, 13 | 1797 | 7106652 | |