siRNA Details
| Gene Symbol: Pdgfrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011921.1 | NP_001011921.1 | 2 | 225 | ||
| GenBank mRNA | BC086555.1 | - | 2 | 225 | 7305039 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pdgfrl| Gene Symbol: Pdgfrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011921.1 | NP_001011921.1 | 2 | 225 | ||
| GenBank mRNA | BC086555.1 | - | 2 | 225 | 7305039 | |