siRNA Details
| Gene Symbol: Prpf4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011923.1 | NP_001011923.1 | 2 | 157 | ||
| GenBank mRNA | BC085927.1 | - | 2 | 157 | 7110150 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prpf4b| Gene Symbol: Prpf4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011923.1 | NP_001011923.1 | 2 | 157 | ||
| GenBank mRNA | BC085927.1 | - | 2 | 157 | 7110150 | |