siRNA Details
| Gene Symbol: Nup93 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011925.1 | NP_001011925.1 | 15 | 1801 | ||
| XM_008772282.1 | XP_008770504.1 | 13 | 3897 | |||
| GenBank mRNA | BC081925.1 | - | 15 | 1801 | 7113037 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nup93| Gene Symbol: Nup93 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011925.1 | NP_001011925.1 | 15 | 1801 | ||
| XM_008772282.1 | XP_008770504.1 | 13 | 3897 | |||
| GenBank mRNA | BC081925.1 | - | 15 | 1801 | 7113037 | |