siRNA Details
| Gene Symbol: Cnnm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011942.1 | NP_001011942.1 | 1 | 1407 | ||
| XM_006231471.2 | XP_006231533.1 | 1 | 1500 | |||
| XM_008760413.1 | XP_008758635.1 | 1 | 1501 | |||
| GenBank mRNA | BC085930.1 | - | 1 | 1407 | 7110800 | |

Cnnm2
Genomic Map