siRNA Details
| Gene Symbol: Cnnm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011942.1 | NP_001011942.1 | 8 | 2837 | ||
| XM_006231471.2 | XP_006231533.1 | 7 | 2864 | |||
| GenBank mRNA | BC085930.1 | - | 8 | 2837 | 7110800 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cnnm2| Gene Symbol: Cnnm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011942.1 | NP_001011942.1 | 8 | 2837 | ||
| XM_006231471.2 | XP_006231533.1 | 7 | 2864 | |||
| GenBank mRNA | BC085930.1 | - | 8 | 2837 | 7110800 | |