siRNA Details
| Gene Symbol: CC2D1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017721.4 | NP_060191.3 | 2 | 442 | ||
| XM_005259972.2 | XP_005260029.1 | 2 | 390 | |||
| XM_005259973.2 | XP_005260030.1 | 2 | 390 | |||
| XM_005259974.2 | XP_005260031.1 | 2 | 390 | |||
| XM_005259975.2 | XP_005260032.1 | 2 | 390 | |||
| GenBank mRNA | AB097002.1 | - | 2 | 384 | ||
| AF536205.1 | - | 4 | 495 | |||
| AK123155.1 | - | 2 | 382 | |||
| AK315068.1 | - | 2 | 360 | |||
| BC064981.1 | - | 2 | 346 | 6585236 | ||
| BC113968.1 | - | 1 | 71 | |||
| DB065701.1 | - | 2 | 443 | |||

CC2D1A
Genomic Map