siRNA Details
| Gene Symbol: Hexb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011946.1 | NP_001011946.1 | 13 | 1709 | ||
| GenBank mRNA | BC079376.1 | - | 13 | 1709 | 7110247 | |
| FQ221834.1 | - | 8 | 833 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hexb| Gene Symbol: Hexb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011946.1 | NP_001011946.1 | 13 | 1709 | ||
| GenBank mRNA | BC079376.1 | - | 13 | 1709 | 7110247 | |
| FQ221834.1 | - | 8 | 833 | |||