siRNA Details
| Gene Symbol: Ccna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011949.1 | NP_001011949.1 | 5 | 836 | ||
| XM_006232359.2 | XP_006232421.1 | 5 | 798 | |||
| XM_006232360.2 | XP_006232422.1 | 5 | 801 | |||
| XM_006232361.2 | XP_006232423.1 | 5 | 841 | |||
| XM_006232362.2 | XP_006232424.1 | 5 | 802 | |||
| XM_006232363.2 | XP_006232425.1 | 5 | 801 | |||
| XM_006232364.2 | XP_006232426.1 | 5 | 803 | |||
| XM_008760987.1 | XP_008759209.1 | 5 | 891 | |||
| XM_008760988.1 | XP_008759210.1 | 5 | 782 | |||
| GenBank mRNA | BC079234.1 | - | 5 | 836 | 7132591 | |

Ccna1
Genomic Map