siRNA Details
| Gene Symbol: Ccna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011949.1 | NP_001011949.1 | 8 | 1424 | ||
| XM_006232359.2 | XP_006232421.1 | 8 | 1386 | |||
| XM_006232360.2 | XP_006232422.1 | 8 | 1389 | |||
| XM_006232361.2 | XP_006232423.1 | 8 | 1429 | |||
| XM_006232363.2 | XP_006232425.1 | 8 | 1317 | |||
| XM_008760987.1 | XP_008759209.1 | 8 | 1479 | |||
| XM_008760988.1 | XP_008759210.1 | 8 | 1370 | |||
| GenBank mRNA | BC079234.1 | - | 8 | 1424 | 7132591 | |

Ccna1
Genomic Map