siRNA Details
| Gene Symbol: Ell3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011957.1 | NP_001011957.1 | 2, 3 | 217 | ||
| XM_006234831.2 | XP_006234893.1 | 2, 3 | 249 | |||
| XM_006234832.2 | XP_006234894.1 | 2, 3 | 104 | |||
| GenBank mRNA | BC084693.1 | - | 2, 3 | 217 | 7107022 | |

Ell3
Genomic Map