siRNA Details
| Gene Symbol: Zswim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011960.1 | NP_001011960.1 | 8 | 1003 | ||
| XM_006234435.2 | XP_006234497.1 | 8 | 1037 | |||
| GenBank mRNA | BC079431.1 | - | 8 | 1003 | 7114765 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zswim2| Gene Symbol: Zswim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011960.1 | NP_001011960.1 | 8 | 1003 | ||
| XM_006234435.2 | XP_006234497.1 | 8 | 1037 | |||
| GenBank mRNA | BC079431.1 | - | 8 | 1003 | 7114765 | |