siRNA Details
| Gene Symbol: Cercam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011962.1 | NP_001011962.1 | 8 | 889 | ||
| XM_006233768.2 | XP_006233830.1 | 5 | 1557 | |||
| XM_006233769.2 | XP_006233831.1 | 6 | 949 | |||
| XM_006233770.2 | XP_006233832.1 | 8 | 1929 | |||
| XM_006233771.2 | XP_006233833.1 | 4 | 1423 | |||
| GenBank mRNA | BC085782.1 | - | 8 | 889 | 7107449 | |

Cercam
Genomic Map