siRNA Details
| Gene Symbol: Cercam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011962.1 | NP_001011962.1 | 15 | 1982 | ||
| XM_006233770.2 | XP_006233832.1 | 15 | 3022 | |||
| GenBank mRNA | BC085782.1 | - | 15 | 1982 | 7107449 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cercam| Gene Symbol: Cercam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011962.1 | NP_001011962.1 | 15 | 1982 | ||
| XM_006233770.2 | XP_006233832.1 | 15 | 3022 | |||
| GenBank mRNA | BC085782.1 | - | 15 | 1982 | 7107449 | |