siRNA Details
| Gene Symbol: Asb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011963.1 | NP_001011963.1 | 6 | 1482 | ||
| XM_006233912.2 | XP_006233974.1 | 6 | 1450 | |||
| XM_008761641.1 | XP_008759863.1 | 5 | 2216 | |||
| GenBank mRNA | BC079099.1 | - | 6 | 1482 | 7124433 | |

Asb6
Genomic Map