siRNA Details
| Gene Symbol: Stom | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011965.1 | NP_001011965.1 | 7 | 1210 | ||
| GenBank mRNA | BC083895.1 | - | 7 | 1210 | 7132959 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stom| Gene Symbol: Stom | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011965.1 | NP_001011965.1 | 7 | 1210 | ||
| GenBank mRNA | BC083895.1 | - | 7 | 1210 | 7132959 | |