siRNA Details
| Gene Symbol: Strap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011969.1 | NP_001011969.1 | 7 | 858 | ||
| GenBank mRNA | BC083714.1 | - | 7 | 858 | 7190676 | |
| Gene Symbol: LOC100910575 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749864.3 | XP_003749912.2 | 7 | 879 | ||

Strap
Genomic Map