siRNA Details
| Gene Symbol: Strap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011969.1 | NP_001011969.1 | 9 | 1164 | ||
| GenBank mRNA | BC083714.1 | - | 9 | 1164 | 7190676 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Strap| Gene Symbol: Strap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011969.1 | NP_001011969.1 | 9 | 1164 | ||
| GenBank mRNA | BC083714.1 | - | 9 | 1164 | 7190676 | |