siRNA Details
| Gene Symbol: PALMD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017734.4 | NP_060204.1 | 2 | 547 | ||
| GenBank mRNA | AF262379.1 | - | 2 | 478 | ||
| AJ312214.1 | - | 2 | 389 | |||
| AK000278.1 | - | 2 | 309 | |||
| AL390971.1 | - | 2 | 477 | |||
| AL390972.1 | - | 2 | 80 | |||
| AL390973.1 | - | 2 | 273 | |||
| BC012570.1 | - | 2 | 362 | 4245261 | ||
| KU178657.1 | - | 2 | 104 | |||
| KU178658.1 | - | 2 | 104 | |||

PALMD
Genomic Map